Anti-CD99 Mouse Monoclonal Antibody (Biotin) [clone: HI156]

Lieferant: Abnova
MAB15385
ABNOMAB15385EA 402 EUR
ABNOMAB15385
Anti-CD99 Mouse Monoclonal Antibody (Biotin) [clone: HI156]
Antikörper
Mouse monoclonal antibody raised against native human CD99.

Type: Primary
Antigen: CD99
Clonality: Monoclonal
Clone: HI156
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR