Anti-ITGAE Mouse Monoclonal Antibody (FITC) [clone: Ber-ACT8]

Lieferant: Abnova
MAB15387
ABNOMAB15387EA 1040 EUR
ABNOMAB15387
Anti-ITGAE Mouse Monoclonal Antibody (FITC) [clone: Ber-ACT8]
Antikörper
Mouse monoclonal antibody raised against native human ITGAE.

Type: Primary
Antigen: ITGAE
Clonality: Monoclonal
Clone: Ber-ACT8
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR