Anti-ENG Mouse Monoclonal Antibody (CF-Blue) [clone: 2H6F11]

Lieferant: Abnova
MAB15393
ABNOMAB15393EA 423 EUR
ABNOMAB15393
Anti-ENG Mouse Monoclonal Antibody (CF-Blue) [clone: 2H6F11]
Antikörper
Mouse monoclonal antibody raised against recombinant human ENG.

Type: Primary
Antigen: ENG
Clonality: Monoclonal
Clone: 2H6F11
Conjugation: CF-Blue
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR