Anti-IL3RA Mouse Monoclonal Antibody (APC) [clone: AC145]

Lieferant: Abnova
MAB15394
ABNOMAB15394EA 1040 EUR
ABNOMAB15394
Anti-IL3RA Mouse Monoclonal Antibody (APC) [clone: AC145]
Antikörper
Mouse monoclonal antibody raised against human IL3RA.

Type: Primary
Antigen: IL3RA
Clonality: Monoclonal
Clone: AC145
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR