Anti-HLA-DR Mouse Monoclonal Antibody (PE) [clone: GRB-1]

Lieferant: Abnova
MAB15423
ABNOMAB15423EA 402 EUR
ABNOMAB15423
Anti-HLA-DR Mouse Monoclonal Antibody (PE) [clone: GRB-1]
Antikörper
Mouse monoclonal antibody raised against native human HLA-DR.

Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: GRB-1
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR