Anti-HLA-DR Mouse Monoclonal Antibody (PerCP) [clone: GRB-1]
Lieferant: Abnova
MAB15425
ABNOMAB15425EA 523 EUR
ABNOMAB15425
Anti-HLA-DR Mouse Monoclonal Antibody (PerCP) [clone: GRB-1]
Antikörper
Mouse monoclonal antibody raised against native human HLA-DR.
Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: GRB-1
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human