Anti-HLA-DR Mouse Monoclonal Antibody (CF-Blue) [clone: L243]

Lieferant: Abnova
MAB15430
ABNOMAB15430EA 523 EUR
ABNOMAB15430
Anti-HLA-DR Mouse Monoclonal Antibody (CF-Blue) [clone: L243]
Antikörper
Mouse monoclonal antibody raised against native human HLA-DR.

Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: L243
Conjugation: CF-Blue
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR