Anti-CD45.2 Mouse Monoclonal Antibody (CF-Blue) [clone: 104]

Lieferant: Abnova
MAB15444
ABNOMAB15444EA 402 EUR
ABNOMAB15444
Anti-CD45.2 Mouse Monoclonal Antibody (CF-Blue) [clone: 104]
Antikörper
Mouse monoclonal antibody raised against native mouse Cd45.2.

Type: Primary
Antigen: CD45.2
Clonality: Monoclonal
Clone: 104
Conjugation: CF-Blue
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR