Anti-H2-D1 Mouse Monoclonal Antibody [clone: 34-2-12]

Lieferant: Abnova
MAB15446
ABNOMAB15446EA 402 EUR
ABNOMAB15446
Anti-H2-D1 Mouse Monoclonal Antibody [clone: 34-2-12]
Antikörper
Mouse monoclonal antibody raised against native mouse H2-D1.

Type: Primary
Antigen: H2-D1
Clonality: Monoclonal
Clone: 34-2-12
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR