Anti-H2-D1 Mouse Monoclonal Antibody (FITC) [clone: 34-2-12]

Lieferant: Abnova
MAB15447
ABNOMAB15447EA 402 EUR
ABNOMAB15447
Anti-H2-D1 Mouse Monoclonal Antibody (FITC) [clone: 34-2-12]
Antikörper
Mouse monoclonal antibody raised against native mouse H2-D1.

Type: Primary
Antigen: H2-D1
Clonality: Monoclonal
Clone: 34-2-12
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR