Anti-Theophylline Mouse Monoclonal Antibody [clone: 11-58.3.1.2]

Lieferant: Abnova
MAB2229
ABNOMAB2229EA 402 EUR
ABNOMAB2229
Anti-Theophylline Mouse Monoclonal Antibody [clone: 11-58.3.1.2]
Antikörper
Mouse monoclonal antibody raised against Theophylline.

Type: Primary
Antigen: Theophylline
Clonality: Monoclonal
Clone: 11-58.3.1.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR