Anti-Quinidine Monoclonal Antibody [clone: 11-228.6.5]

Lieferant: Abnova
MAB2247
ABNOMAB2247EA 402 EUR
ABNOMAB2247
Anti-Quinidine Monoclonal Antibody [clone: 11-228.6.5]
Antikörper
Mouse monoclonal antibody raised against Quinidine.

Type: Primary
Antigen: Quinidine
Clonality: Monoclonal
Clone: 11-228.6.5
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR