Anti-Quinidine Monoclonal Antibody [clone: 13-406.6.1]

Lieferant: Abnova
MAB2250
ABNOMAB2250EA 402 EUR
ABNOMAB2250
Anti-Quinidine Monoclonal Antibody [clone: 13-406.6.1]
Antikörper
Mouse monoclonal antibody raised against Quinidine.

Type: Primary
Antigen: Quinidine
Clonality: Monoclonal
Clone: 13-406.6.1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR