Anti-Quinidine Monoclonal Antibody [clone: 14-504.5]
Lieferant: Abnova
MAB2251
ABNOMAB2251EA 402 EUR
ABNOMAB2251
Anti-Quinidine Monoclonal Antibody [clone: 14-504.5]
Antikörper
Mouse monoclonal antibody raised against Quinidine.
Type: Primary
Antigen: Quinidine
Clonality: Monoclonal
Clone: 14-504.5
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity:
Bestellen