Anti-Lidocaine Monoclonal Antibody [clone: 11-222.4]

Lieferant: Abnova
MAB2253
ABNOMAB2253EA 402 EUR
ABNOMAB2253
Anti-Lidocaine Monoclonal Antibody [clone: 11-222.4]
Antikörper
Mouse monoclonal antibody raised against Lidocaine.

Type: Primary
Antigen: Lidocaine
Clonality: Monoclonal
Clone: 11-222.4
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR