Anti-VPA Monoclonal Antibody [clone: 10-115.2]

Lieferant: Abnova
MAB2266
ABNOMAB2266EA 402 EUR
ABNOMAB2266
Anti-VPA Monoclonal Antibody [clone: 10-115.2]
Antikörper
Mouse monoclonal antibody raised against Valproic acid.

Type: Primary
Antigen: VPA
Clonality: Monoclonal
Clone: 10-115.2
Conjugation:
Epitope:
Host:
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR