Anti-Enrofloxacin Monoclonal Antibody [clone: 7F21G1F71]

Lieferant: Abnova
MAB2280
ABNOMAB2280EA 402 EUR
ABNOMAB2280
Anti-Enrofloxacin Monoclonal Antibody [clone: 7F21G1F71]
Antikörper
Mouse monoclonal antibody raised against Enrofloxacin.

Recommended Dilutions: ELISA: 1000-1:2000

Type: Primary
Antigen: Enrofloxacin
Clonality: Monoclonal
Clone: 7F21G1F71
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgM
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR