Anti-NP Mouse Monoclonal Antibody [clone: FLUA-NP 4F1]

Lieferant: Abnova
MAB3326
ABNOMAB3326EA 402 EUR
ABNOMAB3326
Anti-NP Mouse Monoclonal Antibody [clone: FLUA-NP 4F1]
Antikörper
Mouse monoclonal antibody raised against recombinant influenza A Nucleoprotein.

Recommended Dilutions: Western Blot: 2-5 ?g/ml; Immunostaining (2-5 ?g/ml

Type: Primary
Antigen: NP
Clonality: Monoclonal
Clone: FLUA-NP 4F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Virus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR