Anti-HLAA Mouse Monoclonal Antibody [clone: MEM-123]

Lieferant: Abnova
MAB3854
ABNOMAB3854EA 402 EUR
ABNOMAB3854
Anti-HLAA Mouse Monoclonal Antibody [clone: MEM-123]
Antikörper
Mouse monoclonal antibody raised against native HLA Class I.

Type: Primary
Antigen: HLAA
Clonality: Monoclonal
Clone: MEM-123
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Bovine, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR