Anti-LAT Mouse Monoclonal Antibody [clone: LAT-01]

Lieferant: Abnova
MAB3865
ABNOMAB3865EA 402 EUR
ABNOMAB3865
Anti-LAT Mouse Monoclonal Antibody [clone: LAT-01]
Antikörper
Mouse monoclonal antibody raised against partial recombinant LAT.

Type: Primary
Antigen: LAT
Clonality: Monoclonal
Clone: LAT-01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR