Anti-Dinitrophenol (DNP) Mouse Monoclonal Antibody [clone: B136M]

Lieferant: Abnova
MAB3977
ABNOMAB3977EA 1430 EUR
ABNOMAB3977
Anti-Dinitrophenol (DNP) Mouse Monoclonal Antibody [clone: B136M]
Antikörper
Mouse monoclonal antibody raised against Dinitrophenol (DNP).

Recommended Dilutions: Recommend dilutions start at 5 ?g/ml.

Type: Primary
Antigen: Dinitrophenol (DNP)
Clonality: Monoclonal
Clone: B136M
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR