Anti-APOA1 Mouse Monoclonal Antibody [clone: 057-10029]

Lieferant: Abnova
MAB4010
ABNOMAB4010EA 523 EUR
ABNOMAB4010
Anti-APOA1 Mouse Monoclonal Antibody [clone: 057-10029]
Antikörper
Mouse monoclonal antibody raised against APOA1.

Type: Primary
Antigen: APOA1
Clonality: Monoclonal
Clone: 057-10029
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR