Anti-Ferritin Mouse Monoclonal Antibody [clone: 090-10187]

Lieferant: Abnova
MAB4061
ABNOMAB4061EA 523 EUR
ABNOMAB4061
Anti-Ferritin Mouse Monoclonal Antibody [clone: 090-10187]
Antikörper
Mouse monoclonal antibody raised against native Ferritin.

Type: Primary
Antigen: Ferritin
Clonality: Monoclonal
Clone: 090-10187
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR