Anti-CK4, C5, C6, C8, C10, C13, and C18. Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: C-11]

Lieferant: Abnova
MAB4529
ABNOMAB4529EA 402 EUR
ABNOMAB4529
Anti-CK4, C5, C6, C8, C10, C13, and C18. Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: C-11]
Antikörper
Mouse monoclonal antibody raised against native Cytokeratin.

Type: Primary
Antigen: CK4, C5, C6, C8, C10, C13, and C18.
Clonality: Monoclonal
Clone: C-11
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mammals
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR