Anti-CD2 Mouse Monoclonal Antibody (Biotin) [clone: MEM-65]
Lieferant: Abnova
MAB4556
ABNOMAB4556EA 402 EUR
ABNOMAB4556
Anti-CD2 Mouse Monoclonal Antibody (Biotin) [clone: MEM-65]
Antikörper
Mouse monoclonal antibody raised against native CD2.
Recommended Dilutions: Flow Cytometry: 1:1000
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: MEM-65
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human