Anti-HLA-DR+DP Monoclonal Antibody (Biotin) [clone: MEM-136]

Lieferant: Abnova
MAB5069
ABNOMAB5069EA 402 EUR
ABNOMAB5069
Anti-HLA-DR+DP Monoclonal Antibody (Biotin) [clone: MEM-136]
Antikörper
Mouse monoclonal antibody raised against native HLA-DR+DP.

Recommended Dilutions: Flow Cytometry: 1:400

Type: Primary
Antigen: HLA-DR+DP
Clonality: Monoclonal
Clone: MEM-136
Conjugation: Biotin
Epitope:
Host:
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR