Anti-TCRB Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy5.5® ) [clone: H57-597]

Lieferant: Abnova
MAB5768
ABNOMAB5768EA 423 EUR
ABNOMAB5768
Anti-TCRB Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy5.5® ) [clone: H57-597]
Antikörper
Hamster monoclonal antibody raised against native Tcrb.

Recommended Dilutions: Flow Cytometry: 0.2 ?g/10^6 cells

Type: Primary
Antigen: TCRB
Clonality: Monoclonal
Clone: H57-597
Conjugation: PE (Phycoerythrin)/Cy5.5®
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR