Anti-MAPK3 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: SB46b]

Lieferant: Abnova
MAB5900
ABNOMAB5900EA 402 EUR
ABNOMAB5900
Anti-MAPK3 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: SB46b]
Antikörper
Mouse monoclonal antibody raised against recombinant MAPK1/MAPK3.

Type: Primary
Antigen: MAPK3
Clonality: Monoclonal
Clone: SB46b
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR