Anti-8-Hydroxyguanosine Monoclonal Antibody [clone: N45.1]

Lieferant: Abnova
MAB6116
ABNOMAB6116EA 1040 EUR
ABNOMAB6116
Anti-8-Hydroxyguanosine Monoclonal Antibody [clone: N45.1]
Antikörper
Mouse monoclonal antibody raised against 8-OHdG.

Recommended Dilutions: Immunohistochemistry: 1-10 ?g/ml

Type: Primary
Antigen: 8-Hydroxyguanosine
Clonality: Monoclonal
Clone: N45.1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR