Anti-MOMP Mouse Monoclonal Antibody [clone: CL12-707.21]

Lieferant: Abnova
MAB6155
ABNOMAB6155EA 402 EUR
ABNOMAB6155
Anti-MOMP Mouse Monoclonal Antibody [clone: CL12-707.21]
Antikörper
Mouse monoclonal antibody raised against Chlamydia trachomatis MOMP.

Type: Primary
Antigen: MOMP
Clonality: Monoclonal
Clone: CL12-707.21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: C. trachomatis
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR