Anti-Gram Negative Endotoxin Mouse Monoclonal Antibody

Lieferant: Abnova
MAB6173
ABNOMAB6173EA 402 EUR
ABNOMAB6173
Anti-Gram Negative Endotoxin Mouse Monoclonal Antibody
Antikörper
Mouse monoclonal antibody raised against Gram Negative Endotoxin.

Type: Primary
Antigen: Gram Negative Endotoxin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: E. coli
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR