Anti-Aflatoxin Rat Monoclonal Antibody [clone: 2E4]

Lieferant: Abnova
MAB6373
ABNOMAB6373EA 779 EUR
ABNOMAB6373
Anti-Aflatoxin Rat Monoclonal Antibody [clone: 2E4]
Antikörper
Immunogen: Winstar rats were immunized with Aflatoxin M1 conjugated with BSA

  • Recommended Dilutions: ELISA (100ng)

Specificity:This antibody is reactive to Aflatoxin M1 (AFM1) and Aflatoxin B1 (AFB1), but not reactive to BSA and other irrelevant antigens by ELISA.

Type: Primary
Antigen: Aflatoxin
Clonality: Monoclonal
Clone: 2E4
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR