Anti-KLK3 Mouse Monoclonal Antibody [clone: B729M]

Lieferant: Abnova
MAB6730
ABNOMAB6730EA 779 EUR
ABNOMAB6730
Anti-KLK3 Mouse Monoclonal Antibody [clone: B729M]
Antikörper
Mouse monoclonal antibody raised against PSA (free).

Type: Primary
Antigen: KLK3
Clonality: Monoclonal
Clone: B729M
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR