Anti-KLK3 Mouse Monoclonal Antibody [clone: B731M]

Lieferant: Abnova
MAB6731
ABNOMAB6731EA 779 EUR
ABNOMAB6731
Anti-KLK3 Mouse Monoclonal Antibody [clone: B731M]
Antikörper
Mouse monoclonal antibody raised against PSA (total).

Type: Primary
Antigen: KLK3
Clonality: Monoclonal
Clone: B731M
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR