Anti-KLK3 Mouse Monoclonal Antibody [clone: B728M]

Lieferant: Abnova
MAB6732
ABNOMAB6732EA 779 EUR
ABNOMAB6732
Anti-KLK3 Mouse Monoclonal Antibody [clone: B728M]
Antikörper
Mouse monoclonal antibody raised against PSA.

Type: Primary
Antigen: KLK3
Clonality: Monoclonal
Clone: B728M
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR