Anti-IL1R1 Mouse Monoclonal Antibody [clone: IL1-31-104.1.1]

Lieferant: Abnova
MAB8354
ABNOMAB8354EA 648 EUR
ABNOMAB8354
Anti-IL1R1 Mouse Monoclonal Antibody [clone: IL1-31-104.1.1]
Antikörper
Mouse monoclonal antibody raised against partial IL-1 receptor.

Recommended Dilutions: ELISA: 5 ?g/ml Radio Immunoassay: 20-30 ng/ml; Immunohistochemistry: 10-100 ?g/ml

Type: Primary
Antigen: IL1R1
Clonality: Monoclonal
Clone: IL1-31-104.1.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR