Anti-IgE Mouse Monoclonal Antibody [clone: 4F4]

Lieferant: Abnova
MAB8447
ABNOMAB8447EA 402 EUR
ABNOMAB8447
Anti-IgE Mouse Monoclonal Antibody [clone: 4F4]
Antikörper
Mouse monoclonal antibody raised against native IgE.

Type: Primary
Antigen: IgE
Clonality: Monoclonal
Clone: 4F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR