Anti-CTSC Mouse Monoclonal Antibody [clone: 97a6]

Lieferant: Abnova
MAB8780
ABNOMAB8780EA 523 EUR
ABNOMAB8780
Anti-CTSC Mouse Monoclonal Antibody [clone: 97a6]
Antikörper
Mouse monoclonal antibody raised against full length recombinant CTSC.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CTSC
Clonality: Monoclonal
Clone: 97a6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR