Anti-MAPK Monoclonal Antibody [clone: E32]

Lieferant: Abnova
MAB9746
ABNOMAB9746EA 402 EUR
ABNOMAB9746
Anti-MAPK Monoclonal Antibody [clone: E32]
Antikörper
Mouse monoclonal antibody raised against full length recombinant ERK1.

Recommended Dilutions: Western Blot: 1:1000-1:10000

Type: Primary
Antigen: MAPK
Clonality: Monoclonal
Clone: E32
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR