Anti-GFP Mouse Monoclonal Antibody [clone: GF28R]

Lieferant: Abnova
MAB9749
ABNOMAB9749EA 402 EUR
ABNOMAB9749
Anti-GFP Mouse Monoclonal Antibody [clone: GF28R]
Antikörper
Mouse monoclonal antibody raised against synthetic peptide of GFP tag.

Recommended Dilutions: Western Blot: 1:1000-1:3000; Immunohistochemistry: 1:500-1:2000

Type: Primary
Antigen: GFP
Clonality: Monoclonal
Clone: GF28R
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Jellyfishes
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR