Anti-MSN Mouse Monoclonal Antibody [clone: 30]

Lieferant: Abnova
MAB9965
ABNOMAB9965EA 402 EUR
ABNOMAB9965
Anti-MSN Mouse Monoclonal Antibody [clone: 30]
Antikörper
Mouse monoclonal antibody raised against full length recombinant MSN.

Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: MSN
Clonality: Monoclonal
Clone: 30
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR