Anti-SKP2 Mouse Monoclonal Antibody [clone: 1]

Lieferant: Abnova
MAB9976
ABNOMAB9976EA 402 EUR
ABNOMAB9976
Anti-SKP2 Mouse Monoclonal Antibody [clone: 1]
Antikörper
Mouse monoclonal antibody raised against partial recombinant SKP2.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: SKP2
Clonality: Monoclonal
Clone: 1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR