Anti-HSPD1 Mouse Monoclonal Antibody [clone: 14]

Lieferant: Abnova
MAB9977
ABNOMAB9977EA 402 EUR
ABNOMAB9977
Anti-HSPD1 Mouse Monoclonal Antibody [clone: 14]
Antikörper
Mouse monoclonal antibody raised against full length recombinant HSPD1.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: HSPD1
Clonality: Monoclonal
Clone: 14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR