Anti-CD44 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MEM-263]

Lieferant: Abnova
MAB9988
ABNOMAB9988EA 402 EUR
ABNOMAB9988
Anti-CD44 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MEM-263]
Antikörper
Mouse monoclonal antibody raised against CD44.

Recommended Dilutions: Flow Cytometry: 20 ?l reagent / 100 ?l of whole blood or 10^6 cells

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: MEM-263
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR