Anti-T Polyclonal Antibody

Lieferant: Abnova
PAB10352
ABNOPAB10352EA 779 EUR
ABNOPAB10352
Anti-T Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against Phosphothreonine.

Recommended Dilutions: ELISA: 0.5 ?g/ml; Western Blot: 2.0 ?g/ml; Immunoprecipitation: 10.0 ?g/mg sample)

Type: Primary
Antigen: T
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR