Anti-GABBR1 Polyclonal Antibody

Lieferant: Abnova
PAB12572
ABNOPAB12572EA 779 EUR
ABNOPAB12572
Anti-GABBR1 Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against GABBR1.

Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: GABBR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR