Anti-West Nile Virus Envelope Rabbit Polyclonal Antibody
Lieferant: Abnova
PAB13413
ABNOPAB13413EA 585 EUR
ABNOPAB13413
Anti-West Nile Virus Envelope Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of West Nile Virus Envelope.
Recommended Dilutions: ELISA: 1 ?g/ml
Type: Primary
Antigen: West Nile Virus Envelope
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Virus