Anti-HMGA2-P3 Polyclonal Antibody

Lieferant: Abnova
PAB14453
ABNOPAB14453EA 779 EUR
ABNOPAB14453
Anti-HMGA2-P3 Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of HMGA2-P3.

Type: Primary
Antigen: HMGA2-P3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR