Anti-RRM2B Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB15925
ABNOPAB15925EA 523 EUR
ABNOPAB15925
Anti-RRM2B Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic phosphopeptide of RRM2B.

Recommended Dilutions: ELISA: 1:2000-1:5000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: RRM2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR