Anti-RRM2B Rabbit Polyclonal Antibody

Lieferant: Abnova
PAB16911
ABNOPAB16911EA 779 EUR
ABNOPAB16911
Anti-RRM2B Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of RRM2B .

Recommended Dilutions: ELISA: 1:3000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: RRM2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR